Learn More
Medchemexpress LLC Mesosulfuron-methyl | 208465-21-8 | 98.0% | 5 G
SDP
Catalog No. 5000342823
Encompass_Preferred
Mesosulfuron-methyl is a sulfonylurea herbicide that inhibits acetolactate synthase (ALS) and is used in research for post-emergence control of ryegrass and Avena spp. (wild oat) in wheat fields.
  • Inhibits acetolactate synthase (ALS)
  • Effective for post-emergence control of ryegrass
  • Effective for post-emergence control of Avena spp. (wild oat) in wheat fields
Catalog No. 50-003-42823 Supplier Medchemexpress LLC Supplier No. HY1269875G
Mesosulfuron-methyl is a sulfonylurea herbicide that inhibits acetolactate synthase (ALS) and is used in research for post-emergence control of ryegrass and Avena spp. (wild oat) in wheat fields.
  • Inhibits acetolactate synthase (ALS)
  • Effective for post-emergence control of ryegrass
  • Effective for post-emergence control of Avena spp. (wild oat) in wheat fields