Learn More
Biocare Medical, LLC Mesothelin, 6.0 ml, prediluted
SDP
Supplier:  Biocare Medical, LLC API3175AA
Encompass_Preferred
Mesothelin [MSLN-15C11] is a mouse monoclonal antibody that is intended for laboratory use in the qualitative identification of mesothelin protein by immunohistochemistry (IHC) in formalin-fixed paraffin-embedded (FFPE) human tissues.
Catalog No. NC1816985