Learn More
Abbiotec LLC Mesothelin Antibody Rabbit Polyclonal Antibody, 0.1 mg
SDP
Supplier:  Abbiotec LLC 250519
Encompass
Mesothelin, also called megakaryocyte-potentiating factor (MPF), potentiates megakaryocyte colony formation in vitro. Membrane-anchored forms are involved in cellular adhesion. Antibodies against mesothelin are detected in patients with mesothelioma and ovarian cancer.
Catalog No. NC2754540