Learn More
ACROBiosystems ACROBiosystems Biotinylated Human Mesothelin / MSLN (296-580), Fc Tag, ultra sensitivity (primary amine labeling)
Catalog No. 501957536
Encompass_Preferred
Recombinant Protein;200UG;MABSol Biotinylated Human Mesothelin (296-580), Fc Tag (MSN-H826x) is expressed from human HEK293 cells. It contains AA Glu 296 - Gly 580 (Accession # AAH09272). It is the biotinylated form of Human Mesothelin (296-580), Fc Tag (MSN-H526x).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 59.1 kDa. The protein migrates as 60-68 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7536 Supplier ACROBiosystems Supplier No. MSNH826X200UG
Recombinant Protein;200UG;MABSol Biotinylated Human Mesothelin (296-580), Fc Tag (MSN-H826x) is expressed from human HEK293 cells. It contains AA Glu 296 - Gly 580 (Accession # AAH09272). It is the biotinylated form of Human Mesothelin (296-580), Fc Tag (MSN-H526x).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 59.1 kDa. The protein migrates as 60-68 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.