Learn More
ACROBiosystems ACROBiosystems Human Mesothelin / MSLN (296-580) Protein, His Tag, low endotoxin
Supplier:  ACROBiosystems MSNH522A100UG
Encompass_Preferred
Recombinant Protein;100UG;Human Mesothelin (296-580), His Tag, low endotoxin (MSN-H522a) is expressed from human 293 cells (HEK293). It contains AA Glu 296 - Gly 580 (Accession # AAH09272).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 33.0 kDa. The protein migrates as 38-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Mesothelin (296-580), His Tag, low endotoxin (Cat. No. MSN-H522a) at 0.5 μg/mL (100 μL/well) can bind Anti-MSLN MAb, Human IgG1 with a linear range of 0.2-3 ng/mL (QC tested).
Catalog No. 50-195-7192