Learn More
ACROBiosystems Recombinant Protein;Human Mesothelin / MSLN (296-580) Protein, Fc Tag;HEK293;100UG;MSN,MSLN,Mesothelin,MPF
Supplier: ACROBiosystems MSNH5253100UG
Recombinant Protein;Human Mesothelin / MSLN (296-580) Protein, Fc Tag;100UG;Human Mesothelin, Fc Tag (MSN-H5253) is expressed from human 293 cells (HEK293). It contains AA Glu 296 - Gly 580 (Accession # AAH09272.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 58.6 kDa. The protein migrates as 66-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Mesothelin, Fc Tag (Cat. No. MSN-H5253) at 0.5 μg/mL (100 μL/well) can bind Anti-Human MSLN Mab with a linear range of 0.1-3 ng/mL (QC tested).