Learn More
ACROBiosystems Recombinant Protein;FITC-Labeled Human Mesothelin / MSLN (296-580) Protein, Fc Tag;HEK293;200UG;MSN,MSLN,Mesothelin,MPF
Catalog No. 501657596
Encompass_Preferred
Recombinant Protein;FITC-Labeled Human Mesothelin / MSLN (296-580) Protein, Fc Tag;200UG;FITC-Labeled Human Mesothelin (296-580), Fc Tag (Cat. No. MSN-HF253) is expressed from human HEK293 cells. It contains AA Glu 296 - Gly 580 (Accession # AAH09272.1). It is the FITC labeled form of Human Mesothelin (296-580), Fc Tag (Cat. No. MSN-H5253).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 58.6 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-Human MSLN MAb at 10 μg/mL (100 μL/well) can bind FITC-Labeled Human Mesothelin (296-580), Fc Tag (Cat. No. MSN-HF253) with a linear range of 0.078-0.625 μg/mL (Ex.488 nm/Em.535 nm) (QC tested).;293 cells were transfected with anti-MSLN-scFv and RFP tag. 2e5 of the cells were stained with B. FITC-Labeled Human Mesothelin (296-580), Fc Tag (Cat. No. MSN-HF253, 1 μg/ml) and C. FITC-labeled protein control. A. Non-transfect
Catalog No. 50-165-7596 Supplier ACROBiosystems Supplier No. MSNHF253200UG
Recombinant Protein;FITC-Labeled Human Mesothelin / MSLN (296-580) Protein, Fc Tag;200UG;FITC-Labeled Human Mesothelin (296-580), Fc Tag (Cat. No. MSN-HF253) is expressed from human HEK293 cells. It contains AA Glu 296 - Gly 580 (Accession # AAH09272.1). It is the FITC labeled form of Human Mesothelin (296-580), Fc Tag (Cat. No. MSN-H5253).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 58.6 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-Human MSLN MAb at 10 μg/mL (100 μL/well) can bind FITC-Labeled Human Mesothelin (296-580), Fc Tag (Cat. No. MSN-HF253) with a linear range of 0.078-0.625 μg/mL (Ex.488 nm/Em.535 nm) (QC tested).;293 cells were transfected with anti-MSLN-scFv and RFP tag. 2e5 of the cells were stained with B. FITC-Labeled Human Mesothelin (296-580), Fc Tag (Cat. No. MSN-HF253, 1 μg/ml) and C. FITC-labeled protein control. A. Non-transfect