Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human Mesothelin / MSLN (296-580) Protein, Fc Tag, Avi Tag (Avitag™);HEK293;25UG;MSN,MSLN,Mesothelin,MPF
Supplier:  ACROBiosystems MSNH82F625UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human Mesothelin / MSLN (296-580) Protein, Fc Tag, Avi Tag (Avitag™);25UG;Biotinylated Human Mesothelin, Fc Tag, Avi Tag (MSN-H82F6) is expressed from human 293 cells (HEK293). It contains AA Glu 296 - Gly 580 (Accession # AAH09272.1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 60.4 kDa. The protein migrates as 66-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-Human MSLN MAb at 0.5 μg/mL (100 μL/well) can bind Biotinylated Human Mesothelin, Fc Tag, Avi Tag (Cat. No. MSN-H82F6) with a linear range of 0.2-6 ng/mL (QC tested).
Catalog No. 50-165-7802