Learn More
ACROBiosystems Recombinant Protein;Human Mesothelin / MSLN (37-286) Protein;HEK293;100UG;MSN,MSLN,Mesothelin,MPF
Supplier: ACROBiosystems MSNH5222100UG
Recombinant Protein;Human Mesothelin / MSLN (37-286) Protein;100UG;Human Mesothelin, His Tag (MSN-H5222) is expressed from human 293 cells (HEK293). It contains AA Leu 37 - Arg 286 (Accession # AAH09272).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.6 kDa. The protein migrates as 32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.