Learn More
ACROBiosystems Recombinant Protein;Human Mesothelin / MSLN (37-286) Protein;HEK293;1MG;MSN,MSLN,Mesothelin,MPF
Catalog No. 501658439
Encompass_Preferred
Recombinant Protein;Human Mesothelin / MSLN (37-286) Protein;1MG;Human Mesothelin, His Tag (MSN-H5222) is expressed from human 293 cells (HEK293). It contains AA Leu 37 - Arg 286 (Accession # AAH09272).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.6 kDa. The protein migrates as 32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8439 Supplier ACROBiosystems Supplier No. MSNH52221MG
Recombinant Protein;Human Mesothelin / MSLN (37-286) Protein;1MG;Human Mesothelin, His Tag (MSN-H5222) is expressed from human 293 cells (HEK293). It contains AA Leu 37 - Arg 286 (Accession # AAH09272).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.6 kDa. The protein migrates as 32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.