Learn More
ACROBiosystems Recombinant Protein;Human Mesothelin / MSLN (296-580) Protein, low endotoxin;HEK293;1MG;MSN,MSLN,Mesothelin,MPF
Catalog No. 501657836
Encompass_Preferred
Recombinant Protein;Human Mesothelin / MSLN (296-580) Protein, low endotoxin;1MG;Human Mesothelin, His Tag, low endotoxin (MSN-H522a) is expressed from human 293 cells (HEK293). It contains AA Glu 296 - Gly 580 (Accession # AAH09272).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 33.0 kDa. The protein migrates as 32-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Mesothelin, His Tag, low endotoxin (Cat. No. MSN-H522a) at 2 μg/mL (100 μL/well) can bind Anti-MSLN MAb, Human IgG1 with a linear range of 0.1-3 ng/mL (QC tested).
Catalog No. 50-165-7836 Supplier ACROBiosystems Supplier No. MSNH522A1MG
Recombinant Protein;Human Mesothelin / MSLN (296-580) Protein, low endotoxin;1MG;Human Mesothelin, His Tag, low endotoxin (MSN-H522a) is expressed from human 293 cells (HEK293). It contains AA Glu 296 - Gly 580 (Accession # AAH09272).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 33.0 kDa. The protein migrates as 32-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Mesothelin, His Tag, low endotoxin (Cat. No. MSN-H522a) at 2 μg/mL (100 μL/well) can bind Anti-MSLN MAb, Human IgG1 with a linear range of 0.1-3 ng/mL (QC tested).