Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human Mesothelin / MSLN (296-580), ultra sensitivity (primary amine labeling);HEK293;200UG;MSN,MSLN,Mesothelin,MPF
Supplier:  ACROBiosystems MSNH8223200UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human Mesothelin / MSLN (296-580), ultra sensitivity (primary amine labeling);200UG;MABSol Biotinylated Human Mesothelin / MSLN (296-580) (MSN-H8223) is expressed from human HEK293 cells. It contains AA Glu 296 - Gly 580 (Accession # AAH09272). It is the biotinylated form of Human Mesothelin / MSLN (296-580) Protein ( MSN-H5223).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 33.0 kDa. The protein migrates as 38-42 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-MSLN MAb, Human IgG1 at 2 g/mL (100 L/well) can bind Biotinylated Human Mesothelin, His Tag (Cat. No. MSN-H8223) with a linear range of 0.031-0.25 g/mL (QC tested).;293 cells were transfected with anti-MSLN-scFv and RFP tag. 2e5 of the cells were first stained with B. Biotinylated Human Mesothelin / MSLN (296-580), ultra sensitivity (primary amine labeling) (Cat. No. MSN-H8223, 3 g/ml) and C
Catalog No. 50-165-9160