Learn More
Aviva Systems Biology Corp MESP1 Antibody - C-terminal region (ARP35960_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35960_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MESP1. It was validated on Western Blot. . Alternate names: MGC10676, bHLHc5ApplicationWB. Species reactivity: Cow, Horse, Human
Catalog No. 50-284-480