Learn More
ATCC MES-SA; Uterine Sarcoma; Human (Homo sapiens)
Supplier:  ATCC CRL1976
Encompass
MES-SA is an epithelial cell line that was isolated in 1980 from the uterus of a White, 56-year-old, female patient with uterine sarcoma. This cell line was deposited by BI Sikic.
Catalog No. 50-238-4712