Learn More
Biomatik Corporation Mest Polyclonal Antibody
Supplier:  Biomatik Corporation CAC10748
Encompass_Preferred
Mest Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MEST. The Mest Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Mesoderm-Specific Transcript Homolog Protein (146-335Aa). Applications: ELISA, WB, IHC. Target Synonyms: MEST. Accession Number: Q5EB52. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6829