Learn More
Medchemexpress LLC Mestranol-d2 | 72-33-3 | 99.0% | 1 MG
SDP
Catalog No. 5000407288
Encompass_Preferred
Mestranol-d2 is the deuterium labeled Mestranol. Mestranol is an inactive proagent that becomes biologically active upon conversion to ethinyl estradiol (EE). It acts as an estrogen receptor agonist and combines with a progestin in vivo. It can be used for the research of menopausal hormone or menstrual disorders.
  • Click chemistry reagent for copper-catalyzed azide-alkyne cycloaddition
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes as tracers for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-07288 Supplier Medchemexpress LLC Supplier No. HYB0390S1MG
Mestranol-d2 is the deuterium labeled Mestranol. Mestranol is an inactive proagent that becomes biologically active upon conversion to ethinyl estradiol (EE). It acts as an estrogen receptor agonist and combines with a progestin in vivo. It can be used for the research of menopausal hormone or menstrual disorders.
  • Click chemistry reagent for copper-catalyzed azide-alkyne cycloaddition
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes as tracers for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs