Learn More
Medchemexpress LLC Met-12 | 1242964-46-0 | 99.66% | 1462.65 | 10 MG
SDP
Catalog No. 5000397179
Encompass_Preferred
Met-12 is a small peptide inhibitor known for its action on the Fas receptor. This compound effectively inhibits Fas receptor-mediated photoreceptor cell apoptosis, making it a valuable tool for relevant research. Furthermore, Met-12 has been observed to reduce Caspase activation, contributing to its potential therapeutic applications in fields related to cell death and ocular health.
  • Inhibits Fas receptor-mediated photoreceptor cell apoptosis
  • Reduces Caspase activation
  • Useful in photoreceptor research
Catalog No. 50-003-97179 Supplier Medchemexpress LLC Supplier No. HYP644010MG
Met-12 is a small peptide inhibitor known for its action on the Fas receptor. This compound effectively inhibits Fas receptor-mediated photoreceptor cell apoptosis, making it a valuable tool for relevant research. Furthermore, Met-12 has been observed to reduce Caspase activation, contributing to its potential therapeutic applications in fields related to cell death and ocular health.
  • Inhibits Fas receptor-mediated photoreceptor cell apoptosis
  • Reduces Caspase activation
  • Useful in photoreceptor research