Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Met-12 | 1242964-46-0 | 99.7% | 1462.65 | 5 MG
SDP

Catalog No. 5000399873
Encompass_Preferred

Met-12 is a small peptide inhibitor of the Fas receptor. It can inhibit Fas receptor-mediated photoreceptor cell apoptosis and reduce Caspase activation. Met-12 is suitable for research on photoreceptors.

  • Inhibits Fas receptor-mediated photoreceptor cell apoptosis.
  • Reduces Caspase activation.
  • Suitable for photoreceptor research.
  • Appearance: solid.
  • Color: white to off-white.
  • Ships at room temperature in continental US.
  • Powder storage: -80°C for 2 years; -20°C for 1 year.
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture).
  • Soluble in DMSO: 100 mg/mL.

Catalog No. 50-003-99873 Supplier Medchemexpress LLC Supplier No. HYP64405MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Met-12 is a small peptide inhibitor of the Fas receptor. It can inhibit Fas receptor-mediated photoreceptor cell apoptosis and reduce Caspase activation. Met-12 is suitable for research on photoreceptors.

  • Inhibits Fas receptor-mediated photoreceptor cell apoptosis.
  • Reduces Caspase activation.
  • Suitable for photoreceptor research.
  • Appearance: solid.
  • Color: white to off-white.
  • Ships at room temperature in continental US.
  • Powder storage: -80°C for 2 years; -20°C for 1 year.
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture).
  • Soluble in DMSO: 100 mg/mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.