Learn More
Ace Glass, Inc. MET CLAMP 14/20 6/PK MET CLAMP 14/20 6/PK
SDP
Catalog No. NC0334323
Encompass
14/20 nickel coated steel clamp to snap on/off standard taper joints with ease while not scratching the glass. Clip offers superior chemical and temperature resistance (500¿ C).
Catalog No. NC0334323 Supplier Ace Glass, Inc. Supplier No. 759913
14/20 nickel coated steel clamp to snap on/off standard taper joints with ease while not scratching the glass. Clip offers superior chemical and temperature resistance (500¿ C).