Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human MET (cMET), His Tag Recombinant Protein

Catalog No. PV3143
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV3143 10 μg
PV3588 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PV3143 Supplier Invitrogen™ Supplier No. PV3143
Add to Cart
Edge
Add to Cart

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Cytoplasmic.

MET (cMET) is a receptor-like tyrosine kinase whose dysregulation has been linked to many types of human malignancies. After activation of the ligand, MET interacts with the PI3-kinase subunit PIK3R1, PLCG1, SRC, GRB2, and STAT3. There interactions lead to the activation of signaling cascades including RAS-ERK, PI3, kinase-AKT, and PLCgamma-PKC. MET plays a role in embryonic development including gastrulation, development of muscles and neurons, angiogenesis, and kidney formation. It also plays a role in adults including wound healing, organ regeneration, and tissue remodeling. MET has been linked to cancers including gastric, renal, and breast; therefore, making it a target for cancer therapeutics and diagnostic testing.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08581
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 20mM tris with 0.5mM EDTA, 100mM NaCl, 2mM DTT, 50% glycerol, 0.05% Triton X-100 and no preservative; pH 7.5
Gene ID (Entrez) 4233
Molecular Weight (g/mol) 53.7 kDa
Name Human MET (cMET), His Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AI838057; AUTS9; bHLHe59; CG1705; CG1705-PA; CG1705-PB; cmet; c-met; c-met proto-oncogene; c-Met receptor tyrosine kinase; c-Met/HGF receptor; D249; DFNB97; Dmel\CG1705; Dmel_CG1705; DmMet; EC 2.7.10.1; Hepatocyte growth factor receptor; HGF; HGF receptor; HGF receptor c-Met; HGF/SF receptor; HGFR; HGF-SF receptor; juvenile hormone resistance; met; met proto-oncogene; met proto-oncogene (hepatocyte growth factor receptor); met proto-oncogene tyrosine kinase; MET proto-oncogene, receptor tyrosine kinase; Met/Met1; Met; protein tyrosin kinase; methoprene tolerant; Methoprene-tolerant; Methoprene-tolerant protein; Met-PA; Met-PB; Mett; oncogene MET; Par4; Proto-oncogene c-Met; RCCP2; resistance (1) juvenile H; resistance to juvenile hormone; Rst(1)JH; sc MET; scatter factor receptor; SF receptor; soluble c met; tyrosine-protein kinase Met
Common Name c-Met
Gene Symbol MET
Kinase Family MET (HGFR) Kinase Family
Protein Length 956-1390
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence 956-1390
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical europium phospho-specific activity, Biochemical binding/kinetic
Kinase Group Tyrosine Kinases
Shipping Condition Dry Ice
System Compatibility LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate
Tag Position N-Terminal
Target Entry MET
Target Gene met proto-oncogene
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Catalytic Domain, Cytoplasmic
Form Liquid
Protein Subtype Receptor Tyrosine Kinases
Research Category Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.