Learn More
Eurofins Discoverx Met (D1246N), active, 10 µg. (14-818)
Supplier:  Eurofins Discoverx 14818
Encompass
N-terminal 6xHis-tagged recombinant, human Met amino acids 974-end, containing the mutation D1246N. Met D1246N is a germline mutation that has been identified in patients with hereditary papillary renal carcinomas (HPRC). in vitro studies have demonstrated enhanced kinase activity from this mutant. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9371