Learn More
Medchemexpress LLC Met-Gly-Pro-AMC | 1926163-52-1 | 99.7% | 460.55 | 5 MG
SDP
Catalog No. 5000396733
Encompass_Preferred
Met-Gly-Pro-AMC is a fluorescent peptide substrate of MetAP2. It is intended for research use only.
  • Fluorescent peptide substrate of MetAP2
  • Solid appearance, white to off-white
  • Soluble in H2O at 50 mg/mL (108.57 mM) with ultrasonic
  • Store powder at -80°C for 2 years, or -20°C for 1 year
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month
  • Requires sealed storage, away from moisture
Catalog No. 50-003-96733 Supplier Medchemexpress LLC Supplier No. HYP43445MG
Met-Gly-Pro-AMC is a fluorescent peptide substrate of MetAP2. It is intended for research use only.
  • Fluorescent peptide substrate of MetAP2
  • Solid appearance, white to off-white
  • Soluble in H2O at 50 mg/mL (108.57 mM) with ultrasonic
  • Store powder at -80°C for 2 years, or -20°C for 1 year
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month
  • Requires sealed storage, away from moisture