Learn More
Medchemexpress LLC MET kinase-IN-2 | 2101241-90-9 | 98.4% | 25 MG
SDP
Catalog No. 5000342307
Encompass_Preferred
MET kinase-IN-2 is a potent, selective, orally bioavailable MET kinase inhibitor with an IC50 of 7.4 nM. It exhibits antitumor activity, making it suitable for research in cancer and related disease areas. This product is for research use only.
  • Potent, selective, orally bioavailable MET kinase inhibitor
  • Exhibits antitumor activity
  • Inhibits various cell lines such as U-87 MG, NIH-H460, HT-29, and MKN-45
  • Demonstrates dose-dependent tumor growth inhibition in xenograft models
  • Features favorable pharmacokinetic profiles
Catalog No. 50-003-42307 Supplier Medchemexpress LLC Supplier No. 25MGHY131065
MET kinase-IN-2 is a potent, selective, orally bioavailable MET kinase inhibitor with an IC50 of 7.4 nM. It exhibits antitumor activity, making it suitable for research in cancer and related disease areas. This product is for research use only.
  • Potent, selective, orally bioavailable MET kinase inhibitor
  • Exhibits antitumor activity
  • Inhibits various cell lines such as U-87 MG, NIH-H460, HT-29, and MKN-45
  • Demonstrates dose-dependent tumor growth inhibition in xenograft models
  • Features favorable pharmacokinetic profiles