Learn More
Medchemexpress LLC Met kinase-IN-2 | 2101241-90-9 | 98.4% | 50 MG
SDP
Catalog No. 5000339023
Encompass_Preferred
MET kinase-IN-2 is a potent, selective, and orally bioavailable MET kinase inhibitor with an IC50 of 7.4 nM. It exhibits antitumor activity.
  • Potent and selective MET kinase inhibitor
  • Orally bioavailable
  • Exhibits antitumor activity
Catalog No. 50-003-39023 Supplier Medchemexpress LLC Supplier No. HY13106550MG
MET kinase-IN-2 is a potent, selective, and orally bioavailable MET kinase inhibitor with an IC50 of 7.4 nM. It exhibits antitumor activity.
  • Potent and selective MET kinase inhibitor
  • Orally bioavailable
  • Exhibits antitumor activity