Learn More
Medchemexpress LLC Met kinase-IN-2 | 2101241-90-9 | 98.4% | 562.59 g/mol | C33H27FN4O4 | 5 MG
SDP
Catalog No. 5000262568
Encompass_Preferred
MET kinase-IN-2 is a potent, selective MET (c-Met/HGFR) kinase inhibitor with an enzymatic IC50 of 7.4 nM. The compound shows cellular growth inhibition across multiple cancer cell lines and demonstrates oral antitumor efficacy in xenograft models. It is supplied for preclinical research and is well characterized for purity and molecular properties.
  • Potent MET kinase inhibition (IC50 7.4 nM).
  • Selective profile with additional activity against AXL, Flt4, KDR, Mer, TEK, and TYRO3.
  • Orally bioavailable with demonstrated in vivo antitumor efficacy.
  • Documented cellular activity in multiple cancer cell lines.
  • High purity and characterized molecular weight and formula.
Catalog No. 50-002-62568 Supplier Medchemexpress LLC Supplier No. HY1310655MG
MET kinase-IN-2 is a potent, selective MET (c-Met/HGFR) kinase inhibitor with an enzymatic IC50 of 7.4 nM. The compound shows cellular growth inhibition across multiple cancer cell lines and demonstrates oral antitumor efficacy in xenograft models. It is supplied for preclinical research and is well characterized for purity and molecular properties.
  • Potent MET kinase inhibition (IC50 7.4 nM).
  • Selective profile with additional activity against AXL, Flt4, KDR, Mer, TEK, and TYRO3.
  • Orally bioavailable with demonstrated in vivo antitumor efficacy.
  • Documented cellular activity in multiple cancer cell lines.
  • High purity and characterized molecular weight and formula.