Learn More
Medchemexpress LLC Met kinase-IN-2 | 2101241-90-9 | 98.4% | C33H27FN4O4 | 10MG
SDP
Catalog No. 5000261895
Encompass_Preferred
MET kinase-IN-2 is a potent, selective, orally bioavailable inhibitor of the MET (c-Met/HGFR) receptor tyrosine kinase. It inhibits MET with an IC50 of 7.4 nM, shows antitumor activity in cell-based assays, and has reported activity against several additional kinases and tumor cell lines. Supplied as a solid research chemical for laboratory use.
  • Potent MET kinase inhibition (IC50 = 7.4 nM).
  • Orally bioavailable, enabling in vivo studies.
  • Demonstrated antitumor activity in cell assays.
  • Shows additional kinase activity at higher concentrations.
  • High-purity solid suitable for research applications.
Catalog No. 50-002-61895 Supplier Medchemexpress LLC Supplier No. HY13106510MG
MET kinase-IN-2 is a potent, selective, orally bioavailable inhibitor of the MET (c-Met/HGFR) receptor tyrosine kinase. It inhibits MET with an IC50 of 7.4 nM, shows antitumor activity in cell-based assays, and has reported activity against several additional kinases and tumor cell lines. Supplied as a solid research chemical for laboratory use.
  • Potent MET kinase inhibition (IC50 = 7.4 nM).
  • Orally bioavailable, enabling in vivo studies.
  • Demonstrated antitumor activity in cell assays.
  • Shows additional kinase activity at higher concentrations.
  • High-purity solid suitable for research applications.