Learn More
Eurofins Discoverx Met (M1268T), active, 10 µg. (14-817)
Supplier:  Eurofins Discoverx 14817
Encompass
N-terminal 6xHis-tagged recombinant, human Met amino acids 974-end, containing the mutation M1268T. Met M1268T is a somatic mutation that has been identified in patients with papillary renal carcinomas (HPRC). In vitro studies have demonstrated enhanced kinase activity from this mutant. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9369