Learn More
Medchemexpress LLC Met-RANTES (human) acetate | 99.39% | 7978.07 | 100 UG
SDP
Catalog No. 5000397623
Encompass_Preferred
Met-RANTES (human) acetate is the acetate form of Met-RANTES (human) (HY-P4191). It acts as an antagonist for CCR1 and CCR5. This compound inhibits human MIP-1α and MIP-1β chemokines with IC50 values of 5 nM and 2 nM, respectively. It has been shown to reduce bone destruction and ameliorate adjuvant-induced arthritis (AIA) in rats.
  • Antagonist for CCR1 and CCR5.
  • Inhibits human MIP-1α and MIP-1β chemokines.
  • Reduces bone destruction.
  • Ameliorates adjuvant-induced arthritis (AIA) in rats.
Catalog No. 50-003-97623 Supplier Medchemexpress LLC Supplier No. HYP4191A100UG
Met-RANTES (human) acetate is the acetate form of Met-RANTES (human) (HY-P4191). It acts as an antagonist for CCR1 and CCR5. This compound inhibits human MIP-1α and MIP-1β chemokines with IC50 values of 5 nM and 2 nM, respectively. It has been shown to reduce bone destruction and ameliorate adjuvant-induced arthritis (AIA) in rats.
  • Antagonist for CCR1 and CCR5.
  • Inhibits human MIP-1α and MIP-1β chemokines.
  • Reduces bone destruction.
  • Ameliorates adjuvant-induced arthritis (AIA) in rats.