Learn More
Sino Biological Recombinant Human c-MET/HGFR Protein (D1228Y, GST Tag), Active
Catalog No. 502574193
Encompass_Preferred
Recombinant human MET (D1228Y) (956-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-4193 Supplier Sino Biological Supplier No. M5212D2G50
Recombinant human MET (D1228Y) (956-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.