Learn More
Sino Biological Recombinant Human c-MET/HGFR Protein (T1173I, GST Tag), Active
Catalog No. 502574183
Encompass_Preferred
Recombinant human MET (T1173I) (956-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-4183 Supplier Sino Biological Supplier No. M5212FG50
Recombinant human MET (T1173I) (956-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.