Learn More
Eurofins Discoverx Met (Y1248D), active, 250 µg. (14-816M)
Supplier:  Eurofins Discoverx 14816M
Encompass
N-terminal 6xHis-tagged recombinant, human Met amino acids 974-end, containing the mutation Y1248D. Met Y1248D is a germline mutation that has been identified in patients with hereditary papillary renal carcinomas (HPRC). in vitro studies have demonstrated that this mutation has a demonstrate transforming ability. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9368