Learn More
Eurofins Discoverx Met (Y1248H), active, 250 µg. (14-804M)
Supplier:  Eurofins Discoverx 14804M
Encompass
N-terminal 6xHis-tagged recombinant, human Met amino acids 974-end, containing the mutation Y1248H. Met Y1248H is a somatic mutation that has been identified in patients with papillary renal carcinomas (HPRC). in vitro studies have demonstrated enhanced kinase activity from this mutant. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9364