Learn More
Medchemexpress LLC HY-P1467A 10mg Medchemexpress, [Met5]-Enkephalin, amide (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1467A10MG
Encompass_Preferred
Medchemexpress, HY-P1467A 10mg [Met5]-Enkephalin, amide (TFA) CAS: [Met5]-Enkephalin, amide TFA is an agonist for δ opioid receptors as well as putative ζ (zeta) opioid receptors . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4209