Learn More
Medchemexpress LLC Meta-fexofenadine | 479035-75-1 | MFCD21363595 | 98.6% | 501.66 | C32H39NO4 | 1 MG
SDP
Catalog No. 5000213708
Encompass_Preferred
Meta-fexofenadine is an analytical standard representing an impurity of fexofenadine, provided as an off-white to light-yellow solid for research and analytical applications. It is intended for use in pharmaceutical impurity profiling, analytical method development, and as a reference material; key chemical identifiers include CAS 479035-75-1, molecular formula C32H39NO4, and molecular weight 501.66.
  • Purity 98.6% for reliable analytical reference.
  • Supplied in small package sizes suitable for laboratory use.
  • Solid form with off-white to light yellow appearance.
  • Stable under recommended storage conditions for long-term use.
  • Suitable for impurity profiling and method validation.
Catalog No. 50-002-13708 Supplier Medchemexpress LLC Supplier No. HY1006571MG
Meta-fexofenadine is an analytical standard representing an impurity of fexofenadine, provided as an off-white to light-yellow solid for research and analytical applications. It is intended for use in pharmaceutical impurity profiling, analytical method development, and as a reference material; key chemical identifiers include CAS 479035-75-1, molecular formula C32H39NO4, and molecular weight 501.66.
  • Purity 98.6% for reliable analytical reference.
  • Supplied in small package sizes suitable for laboratory use.
  • Solid form with off-white to light yellow appearance.
  • Stable under recommended storage conditions for long-term use.
  • Suitable for impurity profiling and method validation.