Learn More
Medchemexpress LLC Meta-fexofenadine (meta-MDL-16455) | 479035-75-1 | 98.6% | 501.66 | C32H39NO4 | 5 MG
SDP
Catalog No. 5000213779
Encompass_Preferred
meta-Fexofenadine is an impurity of the antihistamine fexofenadine provided as a research chemical and analytical reference for impurity profiling, method development, and analytical standardization.
  • High purity: 98.63%.
  • Suitable as an analytical standard for impurity profiling and method validation.
  • Available in small milligram pack sizes for laboratory use.
  • Molecular formula C32H39NO4 and molecular weight 501.66 g/mol.
  • Identified by CAS number 479035-75-1 for unambiguous reference.
Catalog No. 50-002-13779 Supplier Medchemexpress LLC Supplier No. HY1006575MG
meta-Fexofenadine is an impurity of the antihistamine fexofenadine provided as a research chemical and analytical reference for impurity profiling, method development, and analytical standardization.
  • High purity: 98.63%.
  • Suitable as an analytical standard for impurity profiling and method validation.
  • Available in small milligram pack sizes for laboratory use.
  • Molecular formula C32H39NO4 and molecular weight 501.66 g/mol.
  • Identified by CAS number 479035-75-1 for unambiguous reference.