Learn More
Medchemexpress LLC Meta-Hoechst trihydrochloride | 2320308-12-9 | 99.2% | 533.88 g·mol⁻¹ | C25H27Cl3N6O | 10 MG
SDP
Catalog No. 5000287372
Encompass_Preferred
HOE-S 785026 trihydrochloride is a blue fluorescent DNA nuclear stain supplied as a solid. It binds to the minor groove of double-stranded DNA and is used for nuclear staining in fluorescence microscopy and cell-based assays. Available in multiple pack sizes and high purity for research applications.
  • Blue fluorescent dye suitable for DNA/nuclear staining.
  • Binds the minor groove of double-stranded DNA for reliable nuclear labeling.
  • High purity for reproducible experimental results.
  • Available in small quantities to support assay development and optimization.
  • Compatible with fluorescence microscopy and cell-based assays.
Catalog No. 50-002-87372 Supplier Medchemexpress LLC Supplier No. HY15561B10MG
HOE-S 785026 trihydrochloride is a blue fluorescent DNA nuclear stain supplied as a solid. It binds to the minor groove of double-stranded DNA and is used for nuclear staining in fluorescence microscopy and cell-based assays. Available in multiple pack sizes and high purity for research applications.
  • Blue fluorescent dye suitable for DNA/nuclear staining.
  • Binds the minor groove of double-stranded DNA for reliable nuclear labeling.
  • High purity for reproducible experimental results.
  • Available in small quantities to support assay development and optimization.
  • Compatible with fluorescence microscopy and cell-based assays.