Learn More
Medchemexpress LLC Meta-phosphoric acid | 37267-86-0 | MFCD00011351 | 500g
SDP
Catalog No. 5000266444
Encompass_Preferred
Metaphosphoric acid 33 5-36 5% (Glacial phosphoric acid 33 5-36 5%) is a precipitant for rapid enzymatic measurement of blood lactate and pyruvate[1]
Catalog No. 50-002-66444 Supplier Medchemexpress LLC Supplier No. HYY1939A500G
Metaphosphoric acid 33 5-36 5% (Glacial phosphoric acid 33 5-36 5%) is a precipitant for rapid enzymatic measurement of blood lactate and pyruvate[1]