Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

meta-Phosphoric Acid, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02003332 Shop All Avantor Products
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-003-332 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 02-003-332 Supplier Avantor Supplier No. 025205

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 37267-86-0
Packaging Poly Bottle
Specific Gravity 1 L = 0.00 kg
Quantity 2.5 kg
Synonym metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, hydrogen phosphonate
InChI Key UEZVMMHDMIWARA-UHFFFAOYSA-N
SMILES OP(=O)=O
Molecular Weight (g/mol) 79.979
PubChem CID 3084658
Grade Meets ACS reagent chemical requirements
Chemical Name or Material meta-Phosphoric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.