Learn More
Cayman Chemical meta-Topolin, Purity: ≥98%, CAS: 75737-38-1, 100 mg
SDP
Catalog No. 502947711
Encompass_Preferred
A cytokinin; increases root and shoot growth of A. polyphylla from 0.5-15 µM; increases soybean mean callus yield at 5 µM
Catalog No. 50-294-7711 Supplier Cayman Chemical Supplier No. 34122100 MG
A cytokinin; increases root and shoot growth of A. polyphylla from 0.5-15 µM; increases soybean mean callus yield at 5 µM