Learn More
Cayman Chemical meta-Topolin, Purity: ≥98%, CAS: 75737-38-1, 50 mg
SDP
Catalog No. 502950384
Encompass_Preferred
A cytokinin; increases root and shoot growth of A. polyphylla from 0.5-15 µM; increases soybean mean callus yield at 5 µM
Catalog No. 50-295-0384 Supplier Cayman Chemical Supplier No. 3412250 MG
A cytokinin; increases root and shoot growth of A. polyphylla from 0.5-15 µM; increases soybean mean callus yield at 5 µM