Learn More
Enzo Life Sciences SCREEN-WELL Metabotropic Glutamatergic ligand library (100µl/well)
SDP
Catalog No. 502010202
Encompass_Preferred
The Metabotropic Glutamatergic Ligands Plate contains 49 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The Metabotropic Glutamatergic Ligands plate is 1 plate of the larger Neurotransmitter Library (BML-2810) which contains over 650 ligands. Long Term Storage: -80°C. Applications: HTS .
Catalog No. 50-201-0202 Supplier Enzo Life Sciences Supplier No. BML28180100
The Metabotropic Glutamatergic Ligands Plate contains 49 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The Metabotropic Glutamatergic Ligands plate is 1 plate of the larger Neurotransmitter Library (BML-2810) which contains over 650 ligands. Long Term Storage: -80°C. Applications: HTS .