Learn More
Braintree Scientific Metabolic Cage Rack, Vertical, 12-Cage Rack
SDP
Catalog No. 501954068
Encompass
Vertical metabolic cage rack Rack for 12 cages 3600**/3700/3701 (Packed flat) Vertical type Dimensions: mm 1240 x 480 x 1900 height Stainless steel, holds up 12 metabolic and diuresis cages for rats and mice. Design permits excellent view off all cages (cages not included). Minimizes movement of individual cages when other cages are serviced or entire rack is moved. Urethane rubber casters in Stainless Steel caster bodies assure easy, quiet handling; two lock for safety. Autoclavable. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4068 Supplier Braintree Scientific Supplier No. 3M12D100
Vertical metabolic cage rack Rack for 12 cages 3600**/3700/3701 (Packed flat) Vertical type Dimensions: mm 1240 x 480 x 1900 height Stainless steel, holds up 12 metabolic and diuresis cages for rats and mice. Design permits excellent view off all cages (cages not included). Minimizes movement of individual cages when other cages are serviced or entire rack is moved. Urethane rubber casters in Stainless Steel caster bodies assure easy, quiet handling; two lock for safety. Autoclavable. **Sold for research purposes only and not for human use**