Learn More
Apexbio Technology LLC DiscoveryProbe™ Metabolism-related Compound Library Plus, 100uL/well(10 mM solution), 96 Well Racks With Screw Cap
SDP
Catalog No. 501998477
Encompass_Preferred
DiscoveryProbe™ Metabolism-related Compound Library Plus is a unique collection of 1205 metabolism-related compounds for metabolism research. Available in stock with overnight delivery and free shipping over $500 Cost-effective and competitive price to save your findings Potent, selective and cell-permeable in inhibiting or activating target molecules Diverse in chemical structure and route of administration (oral/i.m/i.v injection etc.) Detailed files describing potency, selectivity and applications etc. Supported by published data from top peer-reviewed journals Guaranteed high quality with NMR and HPLC validation. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8477 Supplier Apexbio Technology LLC Supplier No. L1032P.1
Add to Cart
Add to Cart
Request a Quote
DiscoveryProbe™ Metabolism-related Compound Library Plus is a unique collection of 1205 metabolism-related compounds for metabolism research. Available in stock with overnight delivery and free shipping over $500 Cost-effective and competitive price to save your findings Potent, selective and cell-permeable in inhibiting or activating target molecules Diverse in chemical structure and route of administration (oral/i.m/i.v injection etc.) Detailed files describing potency, selectivity and applications etc. Supported by published data from top peer-reviewed journals Guaranteed high quality with NMR and HPLC validation. Other sizes are also available. Please inqury us for quote.