Learn More
Cambridge Isotope Laboratories METABOLOMICS AMINO ACID MIX UNLABELED STANDARD - EACH MIX CONTAINS A SOLUTION OF 17 AMINO ACIDS AT 2.5 MMOL/L CONCENTRATION (EXCEPTION L-CYSTINE AT 1.25 MMOL/L) IN 0.1M HCL - 1.2ML
Supplier:  Cambridge Isotope Laboratories MSKA2US1.2
Encompass
METABOLOMICS AMINO ACID MIX UNLABELED STANDARD - EACH MIX CONTAINS A SOLUTION OF 17 AMINO ACIDS AT 2.5 MMOL/L CONCENTRATION (EXCEPTION L-CYSTINE AT 1.25 MMOL/L) IN 0.1M HCL - 1.2ML
Catalog No. NC1584760