Learn More
Cambridge Isotope Laboratories Metabolomics Amino Acid Mix Standard, 1.2 mL
Catalog No. NC1232621
Encompass
Each mix contains a solution of 17 amino acids at 2.5 mmol/L concentration.
Catalog No. NC1232621 Supplier Cambridge Isotope Laboratories Supplier No. MSKA21.2
Each mix contains a solution of 17 amino acids at 2.5 mmol/L concentration.