Learn More
Microsolv Technology Corporation Metabolomics Development Kit, 2.1x50. 1 Cogent Diamond Hydride™ & 1 Cogent Bidentate C18™. 1 PK.
Catalog No. 502270849
Encompass_Preferred
This is an economical way to purchase two HPLC Columns that are often used with LCMS in Metabolomics and many other fields. Each Column has a 2.1mm Internal Diameter and is 50mm Long, packed with 4um particles and 100A pore size and are manufactured with Cogent TYPE-C™ Silica Hydride. Includes the Cogent Diamond Hydride™ and Cogent Bidentate C18™ columns..
This item is not returnable. View return policy
Catalog No. 50-227-0849 Supplier Microsolv Technology Corporation Supplier No. 4300005P2
This is an economical way to purchase two HPLC Columns that are often used with LCMS in Metabolomics and many other fields. Each Column has a 2.1mm Internal Diameter and is 50mm Long, packed with 4um particles and 100A pore size and are manufactured with Cogent TYPE-C™ Silica Hydride. Includes the Cogent Diamond Hydride™ and Cogent Bidentate C18™ columns..