Learn More
Cayman Chemical Metacetamol, Purity: ≥98%, CAS: 621-42-1, 100 g
SDP
Catalog No. 502949989
Encompass_Preferred
A derivative of acetaminophen; not cytotoxic to isolated mouse hepatocytes (LC50 = >10 mM); induces necrosis and depletion of GSH in isolated human hepatocytes at 10 mM; does not induce hepatic necrosis in mice at 900 mg/kg
Catalog No. 50-294-9989 Supplier Cayman Chemical Supplier No. 33404100 G
A derivative of acetaminophen; not cytotoxic to isolated mouse hepatocytes (LC50 = >10 mM); induces necrosis and depletion of GSH in isolated human hepatocytes at 10 mM; does not induce hepatic necrosis in mice at 900 mg/kg