Learn More
Lj Star Inc METACLAMP 1S33
Catalog No. NC3793021
Encompass
METACLAMP > HYGIENIC LIGHT PORT EASIZE: 1",1-1/2" SANITARY CLAMP CONNECTIONRING MATERIAL: DUPLEX STAINLESS STEEL(1.4462)GLASS: BOROSILICATE TO DIN7080SPECIAL DESIGN FOR USL33 LIGHTPER DRAWING: NO 180-L33
Catalog No. NC3793021 Supplier Lj Star Inc Supplier No. METACLAMP 1S33
METACLAMP > HYGIENIC LIGHT PORT EASIZE: 1",1-1/2" SANITARY CLAMP CONNECTIONRING MATERIAL: DUPLEX STAINLESS STEEL(1.4462)GLASS: BOROSILICATE TO DIN7080SPECIAL DESIGN FOR USL33 LIGHTPER DRAWING: NO 180-L33