Learn More
Medchemexpress LLC HY-B1898 50mg Medchemexpress, Metadoxine CAS:74536-44-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB189850MG
Encompass_Preferred
Medchemexpress, HY-B1898 50mg Metadoxine CAS:74536-44-0 Metadoxine blocks adipocyte differentiation in association with inhibition of the protein kinase A-cAMP response element binding protein (PKA-CREB) pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3031