Learn More
Perkin Elmer US LLC Metal Body Nebulizer Venturi O-Ring for AAnalyst
Catalog No. 502113695
Encompass_Preferred
This O-Ring is included in the Corrosion Resistant Metal Body nebulizer for AAnalyst (03030404), Platinum Alloy Nebulizer for AAnalyst (03030299), and the Stainless Steel Body Nebulizer for AAnalyst (03030352).
Catalog No. 50-211-3695 Supplier Perkin Elmer US LLC Supplier No. 09902005
This O-Ring is included in the Corrosion Resistant Metal Body nebulizer for AAnalyst (03030404), Platinum Alloy Nebulizer for AAnalyst (03030299), and the Stainless Steel Body Nebulizer for AAnalyst (03030352).